missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p-Cresyl sulfate potassium, MedChemExpress
MedChemExpress p-Methylphenyl potassium sulfate is a prototype protein-bound uremic toxin.
Brand: Medchem Express HY-111431A
This item is not returnable.
View return policy
Description
p-Methylphenyl potassium sulfate is a prototype protein-bound uremic toxin.Specifications
p-Cresyl sulfate potassium | |
White | |
C7H7KO4S | |
p-Tolyl sulfate potassium | |
O=S(OC1=CC=C(C)C=C1)(O[K])=O | |
226.29 | |
Research | |
Metabolism-protein/nucleotide metabolism |
91978-69-7 | |
Research | |
50 mg | |
DMSO : 125 mg/mL (552.39 mM; ultrasonic and warming and heat to 60°C) ∣H2O : ≥ 100 mg/mL (441.91 mM) | |
226.29 | |
98.0% | |
Solid |
Research purposes only