missing translation for 'onlineSavingsMsg'
Learn More

p-Nitro Blue Tetrazolium Chloride, Molecular Biology Reagent Grade, MP Biomedicals™

Product Code. 11456451
Click to view available options
Quantity:
50 mg
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 298-83-9
Molecular Formula C40H30Cl2N10O6
Molecular Weight (g/mol) 817.644
MDL Number MFCD00012159
InChI Key FSVCQIDHPKZJSO-UHFFFAOYSA-L
Synonym nitrotetrazolium blue chloride, nitro bt, nitro blue tetrazolium, nbt, nitro blue tetrazolium chloride, p-nitrotetrazolium blue, nitrotetrazolium blue, nitroblue tetrazolium chloride, p-nitro blue tetrazolium chloride, nitro tetrazolium bt
PubChem CID 9281
ChEBI CHEBI:9505
IUPAC Name 2-[2-methoxy-4-[3-methoxy-4-[3-(4-nitrophenyl)-5-phenyltetrazol-2-ium-2-yl]phenyl]phenyl]-3-(4-nitrophenyl)-5-phenyltetrazol-2-ium;dichloride
SMILES COC1=C(C=CC(=C1)C2=CC(=C(C=C2)[N+]3=NC(=NN3C4=CC=C(C=C4)[N+](=O)[O-])C5=CC=CC=C5)OC)[N+]6=NC(=NN6C7=CC=C(C=C7)[N+](=O)[O-])C8=CC=CC=C8.[Cl-].[Cl-]

Specifications

Color Yellow
Assay Percent Range 0.981
Quantity 50 mg
Formula Weight 817.6
Grade Molecular Biology Reagent
Physical Form Powder
Chemical Name or Material p-Nitro Blue Tetrazolium Chloride, For HPLC analysis
compliance-icons compliance-icons
Product Identifier
  • p-nitro blue tetrazolium chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Acute toxicity Category 4
Hazard Statement
  • H318-Causes serious eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P264-Wash thoroughly after handling.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : 5,5'-diphenyl-3,3'-bis(4-nitrophenyl)-2,2'-(3,3'-dimethoxybiphenyl-4,4'ylene)ditetrazolium dichlorid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.