missing translation for 'onlineSavingsMsg'
Learn More

p-Terphenyl, Laser Grade 99%, ACROS Organics™

Product Code. 10455281
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 92-94-4
Melting Point 209.0°C to 216.0°C
Color White
Boiling Point 383.0°C
Flash Point 207°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C18H14
Linear Formula C6H5C6H4C6H5
MDL Number MFCD00003061
Quantity 1 g
Beilstein 05, 695
Synonym p-terphenyl, 1,1':4',1-terphenyl, p-diphenylbenzene, p-triphenyl, santowax p, 4-phenylbiphenyl, 4-phenyldiphenyl, para-terphenyl, biphenyl, 4-phenyl, pyrogallol tannin
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in alcohol, ether, benzene and toluene
InChI Key XJKSTNDFUHDPQJ-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)C3=CC=CC=C3
IUPAC Name 1,4-diphenylbenzene
Molecular Weight (g/mol) 230.31
PubChem CID 7115
ChEBI CHEBI:52242
Formula Weight 230.31
Percent Purity 99%
Grade Pure
Physical Form Crystalline Solid
Chemical Name or Material p-Terphenyl, 99%
Show More Show Less
compliance-icons
Product Identifier
  • p-Terphenyl
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'