missing translation for 'onlineSavingsMsg'
Learn More

p-Thiocresol, 98%

Product Code. 11401537
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11401537

Brand: Thermo Scientific Alfa Aesar undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 106-45-6 | C7H8S | 124.201 g/mol

p-Thiocresol is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Thiocresol is used as a pharmaceutical intermediate.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Air sensitive. Store away from oxidizing agents, air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-45-6
Molecular Formula C7H8S
Molecular Weight (g/mol) 124.201
MDL Number MFCD00004851
InChI Key WLHCBQAPPJAULW-UHFFFAOYSA-N
Synonym p-toluenethiol, p-thiocresol, 4-thiocresol, 4-methylthiophenol, p-methylthiophenol, benzenethiol, 4-methyl, 4-toluenethiol, p-tolyl mercaptan, p-thiolcresol, p-tolylthiol
PubChem CID 7811
IUPAC Name 4-methylbenzenethiol
SMILES CC1=CC=C(C=C1)S

Specifications

Melting Point 40°C to 44°C
Boiling Point 195°C
Flash Point 68°C (154°F)
Odor Stench
Quantity 100 g
UN Number UN2811
Beilstein 605761
Sensitivity Air Sensitive
Merck Index 14,9325
Solubility Information Soluble in alcohol. Insoluble in water.
Formula Weight 124.21
Percent Purity 98%
Chemical Name or Material p-Thiocresol
Show More Show Less
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.