Learn More
p-Toluenesulfonic Acid, 12% in Acetic Acid, Thermo Scientific Chemicals
73.60 € - 189.00 €
Specifications
Chemical Name or Material | p-Toluenesulfonic acid |
---|---|
Color | Colorless to Yellow |
Name Note | 12% in Acetic Acid |
CAS | 104-15-4, 64-19-7 |
Boiling Point | 116.0°C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10677391
|
Thermo Scientific Acros
421222500 |
250 g | Glass bottle |
73.60 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10015720
|
Thermo Scientific Acros
421220010 |
1 kg | Glass bottle |
189.00 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
p-Toluenesulfonic acid | |
12% in Acetic Acid | |
116.0°C | |
MFCD00064387 | |
11 to 13% (Alkalimetry) | |
1.0700g/mL | |
p-toluenesulfonic acid, p-toluenesulphonic acid, 4-toluenesulfonic acid, tosic acid, p-tolylsulfonic acid, toluene-4-sulfonic acid, toluenesulfonic acid, tosylic acid, benzenesulfonic acid, 4-methyl, p-methylbenzenesulfonic acid | |
JOXIMZWYDAKGHI-UHFFFAOYSA-N | |
4-methylbenzenesulfonic acid | |
6101 | |
172.2 | |
Glass bottle | |
1.07 | |
Solution |
Colorless to Yellow | |
104-15-4, 64-19-7 | |
C7H8O3S | |
11, 97 | |
01,1172; 04,508; 05,673; 06,597; 07,374; 08,488; 09,471; 11,535; 12,507 | |
15, 9692 | |
Solubility in water: soluble. | |
CC1=CC=C(C=C1)S(=O)(=O)O | |
172.198 | |
CHEBI:27849 | |
Pure | |
1.3825 to 1.3845 | |
41°C |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
GHS P Statement:
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Keep container tightly closed.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 203-180-0
missing translation for 'rtecsNumber' : XT6300000
missing translation for 'tsca' : TSCA
- p-Toluenesulfonic acid, 12% in acetic acid
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 A
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only