missing translation for 'onlineSavingsMsg'
Learn More

p-Toluenesulfonic acid monohydrate, 99%, extra pure, Thermo Scientific Chemicals

26.43 € - 306.00 €

Chemical Identifiers

CAS 6192-52-5
Molecular Formula C7H10O4S
Molecular Weight (g/mol) 190.213
MDL Number MFCD00142137
InChI Key KJIFKLIQANRMOU-UHFFFAOYSA-N
Synonym p-toluenesulfonic acid monohydrate, 4-methylbenzenesulfonic acid hydrate, 4-methylbenzenesulfonic acid monohydrate, p-toluene sulfonic acid monohydrate, unii-3bto78gaff, benzenesulfonic acid, 4-methyl-, monohydrate, ptoluenesulfonic acid, p-toluenesulfonic acid, monohydrate, 3bto78gaff, 4-methylbenzene-1-sulfonic acid hydrate
PubChem CID 521998
IUPAC Name 4-methylbenzenesulfonic acid;hydrate
SMILES CC1=CC=C(C=C1)S(=O)(=O)O.O
View More Specs

Products 5
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10396780
View Documents Promotion Details
Thermo Scientific Acros
139020050
5 g Glass Bottle
26.43 €
5g
Please to purchase this item. Need a web account? Register with us today!
10455993
View Documents Promotion Details
Thermo Scientific Acros
139021000
100 g Plastic bottle
37.29 €
100g
Please to purchase this item. Need a web account? Register with us today!
10356830
View Documents Promotion Details
Thermo Scientific Acros
139025000
500 g Plastic bottle
90.55 €
500g
Please to purchase this item. Need a web account? Register with us today!
10380004
View Documents Promotion Details
Thermo Scientific
139020025
2.5 kg Plastic bottle
112.56 €
2.50kg
Please to purchase this item. Need a web account? Register with us today!
11924561
View Documents Promotion Details
Thermo Scientific Acros
139020100
10 kg Plastic drum
306.00 €
10kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

6192-52-5
190.213
KJIFKLIQANRMOU-UHFFFAOYSA-N
521998
CC1=CC=C(C=C1)S(=O)(=O)O.O
C7H10O4S
MFCD00142137
p-toluenesulfonic acid monohydrate, 4-methylbenzenesulfonic acid hydrate, 4-methylbenzenesulfonic acid monohydrate, p-toluene sulfonic acid monohydrate, unii-3bto78gaff, benzenesulfonic acid, 4-methyl-, monohydrate, ptoluenesulfonic acid, p-toluenesulfonic acid, monohydrate, 3bto78gaff, 4-methylbenzene-1-sulfonic acid hydrate
4-methylbenzenesulfonic acid;hydrate

Specifications

6192-52-5
99%
CH3C6H4SO3H·H2O
01,1172; 04,508; 05,673; 06,597; 07,374; 08,488; 09,471; 11,535; 12,507
Solubility in water: soluble. Other solubilities: soluble in alcohol, ether and chloroform
CC1=CC=C(C=C1)S(=O)(=O)O.O
190.213
190.22
Extra Pure
p-Toluenesulfonic acid monohydrate, >98.5% (total TSA)
100.0°C to 108.0°C
C7H10O4S
MFCD00142137
p-toluenesulfonic acid monohydrate, 4-methylbenzenesulfonic acid hydrate, 4-methylbenzenesulfonic acid monohydrate, p-toluene sulfonic acid monohydrate, unii-3bto78gaff, benzenesulfonic acid, 4-methyl-, monohydrate, ptoluenesulfonic acid, p-toluenesulfonic acid, monohydrate, 3bto78gaff, 4-methylbenzene-1-sulfonic acid hydrate
KJIFKLIQANRMOU-UHFFFAOYSA-N
4-methylbenzenesulfonic acid;hydrate
521998
99%
180°C
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

missing translation for 'rtecsNumber' : DB7164000

Product Identifier
  • p-Toluenesulfonic acid monohydrate

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only