missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p-Toluenesulfonylmethyl isocyanide, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
p-Toluenesulfonylmethyl isocyanide is a synthetic reagent used in the synthesis of triplet drugs with the 1,3,5-trioxazatriquinane skeleton, pyrroles, oxazoles and in imidazoles. It serves as an isonitrile component in a diastereoselective Passerini reaction. Furthermore, it is used in the Van Leusen reaction to convert aldehydes to nitriles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
p-Toluenesulfonylmethyl isocyanide is a synthetic reagent used in the synthesis of triplet drugs with the 1,3,5-trioxazatriquinane skeleton, pyrroles, oxazoles and in imidazoles. It serves as an isonitrile component in a diastereoselective Passerini reaction. Furthermore, it is used in the Van Leusen reaction to convert aldehydes to nitriles.
Solubility
Soluble in chloroform and methanol. Insoluble in water.
Notes
Moisture, air and light sensitive. Store in cool place. Incompatible with strong bases and strong oxidizing agents.
p-Toluenesulfonylmethyl isocyanide is a synthetic reagent used in the synthesis of triplet drugs with the 1,3,5-trioxazatriquinane skeleton, pyrroles, oxazoles and in imidazoles. It serves as an isonitrile component in a diastereoselective Passerini reaction. Furthermore, it is used in the Van Leusen reaction to convert aldehydes to nitriles.
Solubility
Soluble in chloroform and methanol. Insoluble in water.
Notes
Moisture, air and light sensitive. Store in cool place. Incompatible with strong bases and strong oxidizing agents.
Specifications
Specifications
Melting Point | 109°C to 114°C |
Quantity | 100 g |
UN Number | UN2811 |
Beilstein | 3592382 |
Sensitivity | Moisture sensitive |
Solubility Information | Soluble in chloroform and methanol. Insoluble in water. |
Formula Weight | 195.24 |
Percent Purity | ≥97% |
Chemical Name or Material | p-Toluenesulfonylmethyl isocyanide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'