missing translation for 'onlineSavingsMsg'
Learn More

p-Toluoyl chloride, 99%, Thermo Scientific Chemicals

35.28 € - 365.82 €

Chemical Identifiers

CAS 874-60-2
Molecular Formula C8H7ClO
Molecular Weight (g/mol) 154.593
MDL Number MFCD00000696
InChI Key NQUVCRCCRXRJCK-UHFFFAOYSA-N
Synonym p-toluoyl chloride, benzoyl chloride, 4-methyl, p-methylbenzoyl chloride, 4-toluoyl chloride, p-toluyl chloride, p-toluic acid chloride, 4-methylbenzoic acid chloride, 4-methylbenzoylchloride, 4-methyl-benzoyl chloride, unii-092e36pwl4
PubChem CID 13405
IUPAC Name 4-methylbenzoyl chloride
SMILES CC1=CC=C(C=C1)C(=O)Cl
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11423067
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A15581.22
100 g
35.28 €
100g
Please to purchase this item. Need a web account? Register with us today!
11433067
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A15581.36
500 g
103.52 €
500g
Please to purchase this item. Need a web account? Register with us today!
11443067
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A15581.0E
2500 g
365.82 €
2500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

p-Toluoyl chloride was used to prepare antigen, Tolyl-amido-human serum albumin. It is used as a reagent to synthesize (+)-Yashabushitriol, derivatives of Desloratadine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Toluoyl chloride was used to prepare antigen, Tolyl-amido-human serum albumin. It is used as a reagent to synthesize (+)-Yashabushitriol, derivatives of Desloratadine.

Solubility
Reacts with water alcohol.

Notes
Store at room temperature. Incompatible with oxidizing agent, acids, bases, active metals, alcohols, amines.
Specifications

Chemical Identifiers

874-60-2
154.593
NQUVCRCCRXRJCK-UHFFFAOYSA-N
13405
CC1=CC=C(C=C1)C(=O)Cl
C8H7ClO
MFCD00000696
p-toluoyl chloride, benzoyl chloride, 4-methyl, p-methylbenzoyl chloride, 4-toluoyl chloride, p-toluyl chloride, p-toluic acid chloride, 4-methylbenzoic acid chloride, 4-methylbenzoylchloride, 4-methyl-benzoyl chloride, unii-092e36pwl4
4-methylbenzoyl chloride

Specifications

874-60-2
1.169
82°C (179°F)
C8H7ClO
MFCD00000696
471492
p-toluoyl chloride, benzoyl chloride, 4-methyl, p-methylbenzoyl chloride, 4-toluoyl chloride, p-toluyl chloride, p-toluic acid chloride, 4-methylbenzoic acid chloride, 4-methylbenzoylchloride, 4-methyl-benzoyl chloride, unii-092e36pwl4
NQUVCRCCRXRJCK-UHFFFAOYSA-N
4-methylbenzoyl chloride
13405
99%
p-Toluoyl Chloride
°C to 2°C
225°C to 227°C
99%
1.553
UN3265
Moisture sensitive
Reacts with water alcohol.
CC1=CC=C(C=C1)C(=O)Cl
154.593
154.6
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H227

P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H227-H314

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

missing translation for 'einecsNumber' : 212-864-8

missing translation for 'rtecsNumber' : XV1660000

missing translation for 'tsca' : Yes

missing translation for 'storageNote' : Moisture sensitive

Recommended Storage : Ambient temperatures

Product Identifier
  • p-Toluoyl chloride

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : 4-methyl-Benzoyl chloride

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only