missing translation for 'onlineSavingsMsg'
Learn More
p-Tolylmagnesium bromide, approx. 0.5M solution in diethyl ether, AcroSeal™, Thermo Scientific Chemicals Product Code.: 10433464

p-Tolylmagnesium bromide, approx. 0.5M solution in diethyl ether, AcroSeal™, Thermo Scientific Chemicals

Product Code. 10433464
100 mL, AcroSeal™ Glass bottle
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 

Product Code. 10433464

Brand: Thermo Scientific Acros 426761000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 

p-Tolylmagnesium bromide, approx. 0.5M solution in diethyl ether, AcroSeal, Acros Organics, Quantity: 800mL, Packaging: AcroSeal Glass bottle, Molecular Weight: 195.34, CAS: 4294-57-9, 60-29-7, Density: 0.8660g/mL, Flash Point: -40 deg.C, Formula Weight: 195.34

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4294-57-9, 60-29-7
Molecular Formula C7H7BrMg
Molecular Weight (g/mol) 195.34
MDL Number MFCD00000040
InChI Key ZRJNGFJIBZKXTP-UHFFFAOYSA-M
Synonym p-tolylmagnesium bromide, magnesium, bromo 4-methylphenyl, 4-methylphenylmagnesium bromide, bromo 4-methylphenyl magnesium, 4-tolylmagnesium bromide, 4-toluylmagnesium bromide, p-tolyl-magnesium bromide, 4-tolyl magnesium bromide, 4-methylphenylmagnesiumbromide, p-methylphenylmagnesium bromide, grignard reagentShow MoreShow Less
PubChem CID 3644322
IUPAC Name magnesium;methylbenzene;bromide
SMILES CC1=CC=C([Mg]Br)C=C1

Specifications

Density 0.8660g/mL
Flash Point -40°C
Packaging AcroSeal™ Glass bottle
Linear Formula CH3C6H4MgBr
Quantity 100 mL
Specific Gravity 0.866
Formula Weight 195.34
Chemical Name or Material p-Tolylmagnesium bromide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0436951 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number426768000,426761000
Lot NumberA0453787 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number426768000,426761000
Lot NumberA0451454 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number426768000,426761000
Lot NumberA0444421 Certificate TypeCertificate of Analysis Date27/02/2023 Catalog Number426768000,426761000
Lot NumberA0449172 Certificate TypeCertificate of Analysis Date10/01/2023 Catalog Number426768000,426761000
compliance-icons compliance-icons compliance-icons
Product Identifier
  • p-Tolylmagnesium bromide, ca. 0.4M(10%) solution in diethyl ether
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H224-Extremely flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P240-Ground/bond container and receiving equipment.
  • P243-Take precautionary measures against static discharge.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.
  • P403+P235-Store in a well-ventilated place. Keep cool.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
p-Tolylmagnesium bromide, approx. 0.5M solution in diethyl ether, AcroSeal™, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.