missing translation for 'onlineSavingsMsg'
Learn More

p-Tolylmagnesium chloride, 2M (31 wt.%) solution in THF, AcroSeal™, Thermo Scientific Chemicals

Product Code. 10186553
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 696-61-7, 109-99-9
Molecular Formula C7H7ClMg
Molecular Weight (g/mol) 150.888
MDL Number MFCD00672097
InChI Key BLMRDOZWELZHDX-UHFFFAOYSA-M
Synonym p-tolylmagnesium chloride, magnesium, chloro 4-methylphenyl, 4-tolylmagnesium chloride, 4-tolyl-magnesium chloride, 4-methylphenylmagnesium chloride, ybpfqofspmwgka-uhfffaoysa-m, p-tolylmagnesium chloride solution, 1.0 m in thf
PubChem CID 11829629
IUPAC Name magnesium;methylbenzene;chloride
SMILES CC1=CC=[C-]C=C1.[Mg+2].[Cl-]

Specifications

Color Undesignated
Linear Formula CH3C6H4MgCl
Concentration 2 to 2.2M
Density 1.0000g/mL
Formula Weight 150.89
Packaging AcroSeal™ Glass bottle
Specific Gravity 1
Quantity 100 mL
Flash Point −17°C
Physical Form Solution
Chemical Name or Material p-Tolylmagnesium chloride, 2M solution in THF
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • p-Tolylmagnesium chloride, 2M (31 wt.%) solution in THF
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'