missing translation for 'onlineSavingsMsg'
Learn More
p-Xylene, 99+%, extra pure, Thermo Scientific Chemicals Product Code.: 10320694

p-Xylene, 99+%, extra pure, Thermo Scientific Chemicals

Code produit. 10320694
2.5 L, Glass Bottle
missing translation for 'orderingAttributeHoverText'
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Règlementaire: Ce produit est soumis à des droits d'accise sur les produits énergétiques conformément à la directive européenne 2003/96/CEE.

Product Code. 10320694

Brand: Thermo Scientific Acros 220210025

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Règlementaire: Ce produit est soumis à des droits d'accise sur les produits énergétiques conformément à la directive européenne 2003/96/CEE.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 106-42-3
Molecular Formula C8H10
Molecular Weight (g/mol) 106.17
MDL Number MFCD00008556
InChI Key URLKBWYHVLBVBO-UHFFFAOYSA-N
Synonym p-xylene, para-xylene, 1,4-dimethylbenzene, p-methyltoluene, p-dimethylbenzene, p-xylol, benzene, 1,4-dimethyl, 4-xylene, chromar, 4-methyltoluene
PubChem CID 7809
ChEBI CHEBI:27417
IUPAC Name 1,4-xylene
SMILES CC1=CC=C(C)C=C1

Spécification

Melting Point 13.0°C
Density 0.8660g/mL
Boiling Point 138.0°C
Quantity 2.5 L
Flash Point 25°C
Infrared Spectrum Authentic
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.4940 to 1.4960
Assay Percent Range 99+%
Residue after Evaporation 0.005% max.
Specific Gravity 0.866
Linear Formula C6H4(CH3)2
UN Number 1307
Viscosity 0.648 mPa.s (20°C)
Beilstein 05, 382
Water 0.05% max. (Coulometric)
Merck Index 15, 10278
Solubility Information Solubility in water: 0.2g/L (20°C). Other solubilities: soluble in alcohol, ether and most organic, solvents
Formula Weight 106.17
Percent Purity ≥99%
Grade Extra Pure
Chemical Name or Material p-Xylene, Extra Pure
Afficher plus Afficher moins
compliance-icons compliance-icons compliance-icons
Identifiant produit
  • p-Xylene
Mention d'avertissement
  • Danger
Catégorie de danger
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Mention de danger
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Conseils de prudence
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit
p-Xylene, 99+%, extra pure, Thermo Scientific Chemicals >

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis