missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p-Xylene, 99%, pure, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 mL
1 L
2.5 L
10 L
Packaging:
Glass Bottle
Plastic Drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 13.0°C |
Color | Colorless to Yellow |
Density | 0.8660g/mL |
Boiling Point | 138.0°C |
Quantity | 10 L |
Flash Point | 25°C |
Infrared Spectrum | Authentic |
Packaging | Plastic Drum |
Physical Form | Liquid |
Refractive Index | 1.4940 to 1.4960 |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 9
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0465360 | Certificate of Analysis | 22/08/2024 | 158775000,158770100,158770025,158770010 |
A0462473 | Certificate of Analysis | 03/05/2024 | 158775000,158770100,158770025,158770010 |
A0456606 | Certificate of Analysis | 28/09/2023 | 158775000,158770100,158770025,158770010 |
A0453118 | Certificate of Analysis | 11/05/2023 | 158775000,158770100,158770025,158770010 |
A0449977 | Certificate of Analysis | 01/02/2023 |
Safety and Handling



Product Identifier
- p-Xylene
- Danger
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Aspiration toxicity Category 1
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H412-Harmful to aquatic life with long lasting effects.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H226-Flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
p-Xylene, 99%, pure, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction