missing translation for 'onlineSavingsMsg'
Learn More
p-Xylene, 99%, Thermo Scientific Chemicals Product Code.: 11408813

p-Xylene, 99%, Thermo Scientific Chemicals

Product Code. 11408813
2500 mL
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Product Code. 11408813

Brand: Thermo Scientific Alfa Aesar A10534.0F

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

p-Xylene is used as a precursor in the production of benzoic, isophthalic, tetraphillic acids and dimethyle esters, which are used in the manufacture of polyester. It acts as an intermediate in plastic and rubber products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Xylene is used as a precursor in the production of benzoic, isophthalic, tetraphillic acids and dimethyle esters, which are used in the manufacture of polyester. It acts as an intermediate in plastic and rubber products.

Solubility
Miscible with alcohol, ether, acetone, benzene and chloroform. Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-42-3
Molecular Formula C8H10
Molecular Weight (g/mol) 106.17
MDL Number MFCD00008556
InChI Key URLKBWYHVLBVBO-UHFFFAOYSA-N
Synonym p-xylene, para-xylene, 1,4-dimethylbenzene, p-methyltoluene, p-dimethylbenzene, p-xylol, benzene, 1,4-dimethyl, 4-xylene, chromar, 4-methyltoluene
PubChem CID 7809
ChEBI CHEBI:27417
IUPAC Name 1,4-xylene
SMILES CC1=CC=C(C)C=C1

Specifications

Melting Point 12°C to 13°C
Density 0.866
Boiling Point 137°C to 138°C
Quantity 2500 mL
Flash Point 27°C (80°F)
Odor Aromatic
Refractive Index 1.495
UN Number UN1307
Beilstein 1901563
Merck Index 14,10081
Solubility Information Miscible with alcohol,ether,acetone,benzene and chloroform. Immiscible with water.
Formula Weight 106.17
Percent Purity 99%
Chemical Name or Material p-Xylene
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10248354 Certificate TypeCertificate of Analysis Date09/07/2024 Catalog NumberA10534.AE,A10534.0F,A10534.AP
Lot NumberR21K024 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog NumberA10534.AE,A10534.0F,A10534.AP
Lot Number10249976 Certificate TypeCertificate of Analysis Date11/03/2024 Catalog NumberA10534.AE,A10534.0F,A10534.AP
Lot Number10245690 Certificate TypeCertificate of Analysis Date19/10/2023 Catalog NumberA10534.AE,A10534.0F,A10534.AP
Lot NumberR15J018 Certificate TypeCertificate of Analysis Date22/05/2023 Catalog NumberA10534.AE,A10534.0F,A10534.AP
compliance-icons compliance-icons compliance-icons
Product Identifier
  • p-Xylene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : p-Xylene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
p-Xylene, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.