Learn More
Fisherbrand™ Magnetic Stirring Bar Restrainers
Fits ergonomically in one hand. Fisherbrand™ Magnetic Stirring Bar Restrainers retains and secures the stirring bar in position.
Brand: Fisherbrand™
30.64 EUR valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : 41
Additional Details : Weight : 0.05940kg
Description
- Eliminate hazard caused with stir bar splashes when decanting mixing vessels
- After the vessel is drained, the confined stirring bar is securely retrieved from the vacant container without contaminating the solution
- Fabricated with simple-to-clean durable polystyrene
- Houses strong magnet that restrains magnetic stirring bars up to 80mm (3 in.) long
Specifications
3 in. | |
Eliminate hazard caused with stir bar splashes when decanting mixing vessels, capture bars in containers and restrains bars up to 3 in. long | |
Stirring bars |
Fabricated with simple-to-clean durable polypropylene | |
Spinbar™ Magnetic Stirring Bar Restrainer | |
8 cm |