Learn More
Fisherbrand™ Stir Bar Box Set, Octahedral
For use in containers with bases that are slightly curved or uneven. Fisherbrand™ Star Bar Box Set, Octahedral is isostatically molded in pure, FDA-approved PTFE plastic.
Brand:  Fisherbrand™
Code : 41
Additional Details : Weight : 0.20340kg
Product Code. 11547772
  • 63.45 € / 1 set
In Stock
to see stock.
Add to basket
Description
Description
  • Guaranteed inert and leak-free
  • Individually polished to stay cleaner for longer
  • Each set contains:
2 each of 13 x 8mm
2 each of 15 x 8mm
2 each of 25 x 10mm
2 each of 38 x 10mm
2 each of 51 x 10mm
2 each of 64 x 10mm
Order Info
Includes
Specifications
Specifications
2 Followers Each: 113mm x 8mm, 16mm x 8mm, 16mm x 10mm, 25mm x 10mm, 38mm x 10mm, 51mm x 10mm, 64mm x 10mm
White