Learn More
Brady™ i3300 Industrial Label Printer UK with Wi-Fi
The BradyPrinter i3300 Industrial Label Printer can print a wide range of sign and labels in monocolor, including labels to identify wires, products, laboratory samples and facilities.
Brand: Brady™ 149556
1257.80 EUR valid until 2024-12-31
Use promo code "21918" to get your promotional price.
Additional Details : Weight : 7.00000kg
Description
This mid-volume industrial label printer is ideal for identification throughout your facility, including everything from wires and panels to safety and facility labels.
- Wi-Fi connectivity allows you to get data more easily from a variety of sources
- Print smart technology allows the printer to print on the first label, even on sleeves and small labels
- Foolproof drop-in label rolls and ribbons snap into place in 20 seconds
- Prints on more than 825 parts and 70 material options for facility-wide identification
- Contact our Brady™ Technical Support Team to schedule a free phone appointment
- Comes with QWERTY keyboard
Specifications
Color LCD Touchscreen | |
9 in. | |
9.5 in. | |
12 in. | |
Not Autoclavable | |
i3300 Printer, UK Power Cord, Brady™ Workstation Basic Suite, USB Cable, Product CD, Cleaning Kit, Printed Quick Start Guide | |
512 MB RAM, 195 MB IFFS | |
101.6 mm/s | |
Ethernet, USB, USB 2.0 | |
10°C to 40°C | |
5.2 kg | |
1 Year |
Thermal Transfer | |
229 mm | |
241 mm | |
305 mm | |
Monochrome | |
Bulgarian, French, Chinese (Simplified), Croatian, Czech, Norwegian, Polish, English, Romanian, Finnish, Spanish, Chinese (Tradditional), Swedish, Turkish, Chinese (Simplified), Croatian, Czech, Danish, Dutch, English, Estonian, Finnish | |
i3300 | |
300 dpi | |
Windows™ 10, Windows™ 7, Windows™ 8, Windows™ Servers 2012, 2012 R2, and 2016 | |
11.5 lb. | |
Barcode and Inventory Labeling, Hazard Identification, Laboratory Labeling | |
110/240 V |