Learn More
Fisherbrand™ accuSpin 21R Refrigerated Microcentrifuge
GREENER_CHOICE
The Fisherbrand accuSpin 21 Microcentrifuges with a 24 x 1.5/2.0 mL rotor and a ClickSeal biocontainment lid that offers exceptional acceleration of up to 21,000 x g in as little as 12 seconds.
Brand:  Fisherbrand™ 75003249
4120.00 EUR valid until 2024-12-31
Use promo code "21658" to get your promotional price.
Product Code. 17616315
  • 5150.00 € / 1 set
Edge
Description
Description
  • Durable and flexible rotors
  • Outstanding corrosion resistance with lightweight engineered polymer rotors
  • Standard rotor runs 24 microcentrifuge tubes in a single row, from 1.5 to 2.0 mL tubes to mini-preps and spin columns
  • ClickSeal™ Biocontainment solution
  • Ergonomic one-click rotor opening and closing, as well as transparent design for added safety and convenience with Thermo Scientific ClickSeal biocontainment lids
  • Quiet, Easy to Use Design
  • Simplified operation with intuitive controls, easy-to-read displays and fast one-click centrifuge lid closure
  • Sleek, lightweight, space-saving design
  • Durable and easy to clean and maintain
  • Quiet performance promotes a safe, stress-free environment
TRUSTED_SUSTAINABILITY
Specifications
Specifications
24 x 1.5/2 mL
21,000 x g
440 mm
24 x 1.5/2 mL rotor with ClickSeal Biocontainment Lid
Yes
14,800 rpm
330 mm
292 mm
230 V
Yes