missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Black 6000 Series Halogen Free Thermal Transfer Printer Ribbons
High density halogen free ribbon, excellent solvent resistance, high heat resistance, and super scratch resistance.
33.00 € - 55.00 €
Product Code | Brand | Width (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Width (Metric) | Price | Quantity & Availability | |||||
15741427
|
Brady™
196516 |
110 mm |
55.00 €
Each |
Estimated Shipment: 19-07-2024
Log in to see stock. |
|||||
15911021
|
Brady™
196515 |
65 mm |
33.00 €
Each |
Estimated Shipment: 19-07-2024
Log in to see stock. |
|||||
Description
- High density halogen free ribbons
- Excellent solvent resistance
- High heat and scratch resistance
- Compatible with Brady™ BBP12 and BBP11 label printers
- Resin ribbon formulation
Specifications
Black | |
Resin | |
70 m |