missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals P505-15, 95%
Click to view available options
Quantity:
100 mg
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
CAS | 1370261-96-3 |
Assay Percent Range | 95% |
Quantity | 100 mg |
Molecular Formula | C19H23N9O |
Formula Weight | 393.45 |
Chemical Name or Material | P505-15 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction