Learn More
P5CS Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 17719-1-AP-150UL
 View more versions of this product
Product Code. 16827664
  • 410.00 € / 150µL
Explore more special offers
Description
Description
This gene is a member of the aldehyde dehydrogenase family and encodes a bifunctional ATP- and NADPH-dependent mitochondrial enzyme with both gamma-glutamyl kinase and gamma-glutamyl phosphate reductase activities. The encoded protein catalyzes the reduction of glutamate to delta1-pyrroline-5-carboxylate, a critical step in the de novo biosynthesis of proline, ornithine and arginine. Mutations in this gene lead to hyperammonemia, hypoornithinemia, hypocitrullinemia, hypoargininemia and hypoprolinemia and may be associated with neurodegeneration, cataracts and connective tissue diseases. Alternatively spliced transcript variants, encoding different isoforms, have been described for this gene.
Specifications
Specifications
P5CS
Polyclonal
Unconjugated
ALDH18A1
ALDH18A1, GK, GPR, GSAS, P5CS, PYCS
Rabbit
Antigen Affinity Chromatography
RUO
361755, 56454, 5832
-20°C
Liquid
Immunoprecipitation, Western Blot
0.2 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P54886, Q9Z110
Aldh18a1
P5CS Fusion Protein Ag12100
150 μL
Primary
Human, Mouse, Rat