Learn More
PAβN dihydrochloride, MedChemExpress
MedChemExpress PAβN dihydrochloride (MC-207110 dihydrochloride) is an efflux pump inhibitor.

62.10 € - 656.10 €
Specifications
Chemical Name or Material | PAβN dihydrochloride |
---|---|
Physical Form | Solid |
CAS | 100929-99-5 |
Color | White |
Purity Grade Notes | Research |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16493368
![]() |
Medchem Express
HY-101444A |
10 mg |
117.45 €
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16489896
![]() |
Medchem Express
HY-101444A |
50 mg |
284.85 €
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16479845
![]() |
Medchem Express
HY-101444A |
250 mg |
656.10 €
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16462736
![]() |
Medchem Express
HY-101444A |
25 mg |
183.60 €
25mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16436097
![]() |
Medchem Express
HY-101444A |
5 mg |
62.10 €
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16414616
![]() |
Medchem Express
HY-101444A |
100 mg |
369.90 €
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PAβN dihydrochloride (MC-207110 dihydrochloride) is an efflux pump inhibitor.Specifications
PAβN dihydrochloride | |
100929-99-5 | |
Research | |
COVID-19-immunoregulation | |
DMSO : 83.33 mg/mL (160.41 mM; Need ultrasonic) ∣H2O : 14.29 mg/mL (27.51 mM; Need ultrasonic) | |
519.47 | |
98.51% |
Solid | |
White | |
C25H32Cl2N6O2 | |
MC-207,110 dihydrochloride Phe-Arg-β-naphthylamide dihydrochloride | |
O=C([C@H](CCCNC(N)=N)NC([C@H](CC1=CC=CC=C1)N)=O)NC2=CC(C=CC=C3)=C3C=C2.Cl.Cl | |
519.47 | |
Research |
Safety and Handling
missing translation for 'shelfLife' : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Recommended Storage : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.