Learn More
Thermo Scientific™ μPAC™ ESI-MS Accessories
Use μPAC™ ESI-MS Accessories for fast, reproducible, and low dispersion connection of μPAC HPLC columns to Nanospray Flex™ and EASY-Spray™ ion sources.
Brand:  Thermo Scientific™ EMI-FLEXIONB
 View more versions of this product
Product Code. 17827893
  • 1687.74 € / Each
Estimated Shipment: 07-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Use Thermo Scientific™ μPAC™ ESI-MS Accessories to facilitate reproducible, low dispersion connections between μPAC HPLC columns and low-flow mass spectrometer ion sources.
The μPAC Flex iON Connect ESI-MS Interface is compatible with 360 μm OD electrospray emitters and Thermo Scientific™ Nanospray Flex™ Ion Sources via one-piece fingertight fittings.
The μPAC EASY-Spray Emitter is equipped with a stainless steel 50 μm through-bore union. Achieve high resolution separations in low-flow applications when using this emitter to connect a μPAC HPLC column and a transfer line with a Thermo Scientific™ EASY-Spray™ source.
The μPAC Flex iON Connect ESI-MS Interface consists of two separate units:
  • A docking unit that can be slid onto the Nanospray Flex Ion Source's metal rod and receives both a grounding and a high voltage cable
  • A spray unit compatible with the outlet fittings of μPAC columns on one side and compatible with any 360 μm OD emitter on the other side.
The μPAC EASY-Spray emitter is designed for easy connection to an EASY-Spray MS Source. It contains a pre-assembled EASY-Spray emitter with a transfer line and grounding union.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
μPAC™ Flex iON Connect ESI-MS Interface
Nanospray Flex™ Ion Source