missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ μPAC™ Neo Low-Load Trapping Columns

Combine μPAC™ Neo trapping columns with μPAC™ Neo low-load HPLC columns for optimal performance of bottom-up proteomics experiments.

Brand:  Thermo Scientific™ COL-TRPLOLONEOB2

960.22 EUR valid until 2025-03-14
Use promo code "24142" to get your promotional price.


Product Code. 17807893

  • 1072.38 € / Pack of 2
Estimated Shipment: 28-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Combine Thermo Scientific™ μPAC™ Neo low-load trapping columns with μPAC Neo low-load HPLC columns for optimal performance of bottom-up proteomics experiments. Specifically designed for single-cell proteomics applications, these trapping columns support the use of TMT-labeled samples, with up 10 ng of sample load.

Sample Preconcentration - Perform sample preconcentration in your preferred workflow, with both back flush and forward flush mobile phase direction capabilities.

Decreased Sample Loading Times - Shorten low-flow HPLC analysis run times by injecting your samples at 5-10 μL/min.

Sample Clean-up - Effectively remove salts, reagents, and contaminants that have no affinity for hydrophobic stationary phases from your samples, increasing your analytical column's lifetime and improving spectral quality and detection sensitivity.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

5 to 20 μL/min
1.5 to 7
40°C
Silicon Chip
Yes
10 mm
2.5 μm
18 μm
5800 psi (400 bar)
59%
C8
Micro Pillar Array
Reversed Phase
2 mm
5 μm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.