missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ Packaging Bags with Closure

Designed with zipper closure for storing, collecting, protecting and shipping. Buerkle™ Packaging Bags with Closure are suited for specimens, samples and sales products.

Brand:  Buerkle™ 2348-0008

24.54 EUR valid until 2024-12-31
Use promo code "21745" to get your promotional price.


 View more versions of this product

Product Code. 10765183

  • 30.75 € / Pack of 100
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Bags with a patented zipper closure.  Contents remain dust-free. The film is waterproof and has good tear-resistance.
  • LDPE, crystal-clear
  • 0.05mm strong film
  • Suitable for use with foodstuff
Specifications

Specifications

2500 mL
For Storing, Collecting, Protecting and Shipping of Specimens, Samples and Sales Products and Foodstuffs
0.050 mm
300 mm
Clear
LDPE
100/Pk.
200 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers