Learn More
Buerkle™ Packaging Bags with Closure
Designed with zipper closure for storing, collecting, protecting and shipping. Buerkle™ Packaging Bags with Closure are suited for specimens, samples and sales products.
9.25 € - 68.85 €
Specifications
Color | Clear |
---|---|
Material | LDPE |
Thickness (Metric) | 0.050 mm |
Quantity | 100/Pk. |
Product Code | Brand | Capacity (Metric) | Length (Metric) | Width (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Length (Metric) | Width (Metric) | Price | Quantity & Availability | |||
15188745
|
Buerkle™
2348-0003 |
140 mL | 120 mm | 80 mm |
11.10 €
1 pack |
In Stock
Log in to see stock. |
|||
15524916
|
Buerkle™
2348-0001 |
100 mL | 130 mm | 60 mm |
9.75 €
Pack of 100 |
In Stock
Log in to see stock. |
|||
15534916
|
Buerkle™
2348-0002 |
80 mL | 100 mm | 70 mm |
9.25 €
Pack of 100 |
In Stock
Log in to see stock. |
|||
10198941
|
Buerkle™
2348-0004 |
300 mL | 150 mm | 100 mm |
13.50 €
Pack of 100 |
In Stock
Log in to see stock. |
|||
10742001
|
Buerkle™
2348-0005 |
400 mL | 170 mm | 120 mm |
17.15 €
Pack of 100 |
In Stock
Log in to see stock. |
|||
10441151
|
Buerkle™
2348-0006 |
1000 mL | 220 mm | 160 mm |
22.60 €
Pack of 100 |
In Stock
Log in to see stock. |
|||
10451151
|
Buerkle™
2348-0007 |
1500 mL | 250 mm | 180 mm |
24.70 €
Pack of 100 |
In Stock
Log in to see stock. |
|||
10765183
|
Buerkle™
2348-0008 |
2500 mL | 300 mm | 200 mm |
30.75 €
Pack of 100 |
In Stock
Log in to see stock. |
|||
10550562
|
Buerkle™
2348-0009 |
4500 mL | 350 mm | 250 mm |
68.85 €
Pack of 100 |
In Stock
Log in to see stock. |
|||
10061813
|
Buerkle™
2348-0010 |
6500 mL | 400 mm | 300 mm |
53.80 €
Pack of 100 |
In Stock
Log in to see stock. |
|||
Description
Bags with a patented zipper closure. Contents remain dust-free. The film is waterproof and has good tear-resistance.- LDPE, crystal-clear
- 0.05mm strong film
- Suitable for use with foodstuff
Specifications
Clear | |
LDPE | |
0.050 mm | |
100/Pk. |