missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Paclitaxel, 99+%

Product Code. 10516261
1 mg
Click to view available options
Quantity:
1 mg
5 mg
25 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Paclitaxel, also known as Taxol, CAS # 33069-62-4 is a chemotherapeutic drug that acts through stabilizing microtubule assembly and arresting cell cycle progression.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Paclitaxel is a naturally ocurring compound originally isolated from the tree T. brevifolia
  • Paclitaxel binds tubulin and stabilizes microtubule formation, preventing disassembly
  • Paclitaxel is used as a drug to arrest cells at the mitotic checkpoint and block cell cycle progression, thus inducing death of cancer cells

Applications

  • Paclitaxel can be used in preclinical laboratory research on diseases such as cancer, heart disease, inflammatory disorders, and hepatic fibrosis
  • Paclitaxel can be used as a tool in cell biology to study mitotic checkpoint arrest conditions and downstream signaling cascades
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33069-62-4
Molecular Formula C47H51NO14
Molecular Weight (g/mol) 853.92
MDL Number MFCD00869953
InChI Key RCINICONZNJXQF-VAZQATRQSA-N
Synonym paclitaxel, taxol, taxol a, abraxane, paxene, paxceed, plaxicel, yewtaxan, onxol, ebetaxel
PubChem CID 133640187
IUPAC Name (1S,2S,4S,7R,9S,10S,12R,15S)-4,12-bis(acetyloxy)-1,9-dihydroxy-15-{[(2R,3S)-2-hydroxy-3-phenyl-3-(phenylformamido)propanoyl]oxy}-10,14,17,17-tetramethyl-11-oxo-6-oxatetracyclo[11.3.1.0³,¹⁰.0⁴,⁷]heptadec-13-en-2-yl benzoate
SMILES CC(=O)O[C@@H]1C2=C(C)[C@H](C[C@@](O)([C@@H](OC(=O)C3=CC=CC=C3)C3[C@@]4(CO[C@@H]4C[C@H](O)[C@@]3(C)C1=O)OC(C)=O)C2(C)C)OC(=O)[C@H](O)[C@@H](NC(=O)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 213.0°C to 216.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99% min. (HPLC)
Quantity 1 mg
Merck Index 15, 7081
Solubility Information Solubility in water: sparingly soluble. Other solubilities: soluble in methanol,dmso,acetonitrile
Formula Weight 853.9
Percent Purity 99+%
Physical Form Powder
Chemical Name or Material Paclitaxel, 99+%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Paclitaxel
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 4
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H361-Suspected of damaging fertility or the unborn child.
  • H413-May cause long lasting harmful effects to aquatic life.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Paclitaxel, 99+% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.