missing translation for 'onlineSavingsMsg'
Learn More

PAF (C16), Tocris Bioscience™

Product Code. 30001331
Click to view available options
Quantity:
1 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Endogenous platelet-activating factor (PAF)

Endogenous platelet-activating factor (PAF) and ligand for PAF receptors. Produced by inflammatory cells and a potent chemoattractant for polymorphonuclear neutrophils. Induces increased vascular permeability.

Specifications

CAS 74389-68-7
Molecular Formula C26H54NO7P
MDL Number MFCD00043214
Quantity 1 mg
Synonym platelet activating factor, platelet-activating factor, c16-paf, hag-pc, aprl, hexadecyl-paf-acether, paf c16, 1-o-hexadecyl-2-acetyl-sn-glycero-3-phosphocholine, 1-o-hexadecyl-platelet-activating factor, platelet-activating factor c16
InChI Key HVAUUPRFYPCOCA-AREMUKBSSA-N
SMILES CCCCCCCCCCCCCCCCOC[C@H](COP([O-])(=O)OCC[N+](C)(C)C)OC(C)=O
IUPAC Name (2-{[(2R)-2-(acetyloxy)-3-(hexadecyloxy)propyl phosphonato]oxy}ethyl)trimethylazanium
Molecular Weight (g/mol) 523.69
PubChem CID 108156
ChEBI CHEBI:44811
Formula Weight 523.68
Percent Purity >98%
Chemical Name or Material PAF (C16)
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.