missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ PageRuler™ Prestained Protein Ladder, 10 to 180 kDa

Thermo Scientific PageRuler Prestained Protein Ladder is a mixture of 10 blue-, orange-, and green-stained proteins (10 to 180 kDa) for use as size standards in protein electrophoresis (SDS-PAGE) and western blotting.

Brand:  Thermo Scientific™ 26617

455.82 EUR valid until 2024-12-20
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 11822124

  • 642.00 € / Pack of 10
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Thermo Scientific PageRuler Prestained Protein Ladder is a mixture of 10 blue-, orange-, and green-stained proteins (10 to 180 kDa) for use as size standards in protein electrophoresis (SDS-PAGE) and western blotting. The protein ladder is supplied in a ready-to-use format for direct loading onto gels; no need to heat, reduce, or add sample buffer prior to use.

Applications

  • Monitoring protein migration during SDS-polyacrylamide gel electrophoresis
  • Monitoring protein transfer onto membranes after western blotting
  • Sizing of proteins on SDS-polyacrylamide gels and western blots
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Contents: ten vials of 250 μL each

Storage buffer: 62.5 mM Tris-H3PO4 (pH 7.5 at 25°C), 1 mM EDTA, 2% SDS, 10 mM DTT, 1 mM NaN3 and 33% glycerol

Storage: Upon receipt store at -20°C

10
Yes
3 colors: Blue, Orange, Green
180, 130, 100, 70, 55, 40, 35, 25, 15, 10 kDa
10 x 250 μL
Western Blotting, SDS-PAGE
Colorimetric, NIR Fluorescence (700 nm), RGB Fluorescence (555 nm)
Protein Ladder
10 to 180 kDa
Bolt™ Bis-Tris Plus Gels, Novex™ Tricine Gels, Novex™ Tris-Glycine Gels, NuPAGE™ Bis-Tris Gels, NuPAGE™ Tris-Acetate Gels, SDS-PAGE Gels
PageRuler™
Approved for shipment on Wet or Dry Ice
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.