Learn More
Thermo Scientific™ PageRuler™ Unstained Broad Range Protein Ladder
A mixture of 11 proteins (5 to 250kDa) for use as size standards in protein electrophoresis (SDS-PAGE) and Western blotting.
99.75 € - 316.00 €
Specifications
Number of Markers | 11 |
---|---|
Product Type | Protein Ladder |
Ready to Load | Yes |
Size Range | 5 to 250 kDa |
Stain Type | Unstained |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11892124
|
Thermo Scientific™
26630 |
2 x 250 μL |
99.75 €
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17116108
|
Thermo Scientific™
26630X4 |
8 x 250 μL |
316.00 €
Pack of 4 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Thermo Scientific™ PageRuler Unstained Broad Range Protein Ladder is a mixture of 11 proteins (5 to 250kDa) for use as size standards in protein electrophoresis (SDS-PAGE) and Western blotting.
The protein MW markers in this ladder resolve into clearly identifiable sharp bands ranging in size from 5kDa to 250kDa when analyzed by SDS-PAGE and stained with coomassie dye or silver. The 100, 50 and 20kDa protein bands are of greater intensity and serve as reference bands. Proteins can be detected on Western blots by staining with Ponceau S, coomassie blue or other protein stains. In addition, the ladder proteins (except for the 5kDa peptide) contain an integral Strep-tag™ II Sequence and may be detected on Western blots using Strep-Tactin™ Conjugates. The PageRuler Unstained Broad Range Protein Ladder is supplied ready-to-use: no heating, dilution or addition of a reducing agent is required before use.Highlights:
Size range – eleven proteins spanning 5 to 250kDa
Ready-to-use – supplied in a loading buffer for direct loading on gels; no need to boil
Sharp bands – excellent accuracy for detection in with coomassie or silver stains
Reference bands – the 100, 50 and 20kDa bands are more intense for easy orientation
Tagged – each protein in the ladder contains an integral Strep-tag™ II Sequence
Quality tested – each lot evaluated by SDS-PAGE and Western blotting
Includes:
Proteins in 62.5mM Tris-H3PO4 (pH 7.5 at 25°C), 1mM EDTA, 2% SDS, 100mM DTT, 1mM NaN3, 0.01% bromophenol blue and 33% glycerol.
Recommended for:
Accurate protein sizing on SDS-polyacrylamide gels and Western blots
Specifications
11 | |
Yes | |
Unstained | |
PageRuler™ |
Protein Ladder | |
5 to 250 kDa | |
250, 150, 100, 70, 50, 40, 30, 20, 15, 10, 5 kDa |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.