missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Palladium on 2-4 mm alumina pellets, 0.5% Pd, Thermo Scientific Chemicals
134.00 € - 1675.00 €
Chemical Identifiers
CAS | 7440-05-3 |
---|---|
Molecular Formula | Pd |
Molecular Weight (g/mol) | 106.42 |
MDL Number | MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745 |
InChI Key | KDLHZDBZIXYQEI-UHFFFAOYSA-N |
Synonym | black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon |
PubChem CID | 23938 |
ChEBI | CHEBI:33363 |
IUPAC Name | palladium |
SMILES | [Pd] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10431241
|
Thermo Scientific Acros
195080500 |
50 g | Glass bottle |
134.00 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10226730
|
Thermo Scientific Acros
195082500 |
250 g | Glass bottle |
459.00 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11926951
|
Thermo Scientific Acros
195080010 |
1 kg | Glass bottle |
1675.00 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-05-3 | |
106.42 | |
KDLHZDBZIXYQEI-UHFFFAOYSA-N | |
23938 | |
palladium |
Pd | |
MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745 | |
black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon | |
CHEBI:33363 | |
[Pd] |
Specifications
Gray | |
700g/L (tapped) | |
MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745 | |
black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon | |
[Pd] | |
106.42 | |
CHEBI:33363 | |
0.5% (Pd) | |
Palladium on 1/8-inch alumina pellets |
7440-05-3 | |
Pd | |
01,778; 04,365; 05,498; 06,443; 07,274; 15,13; 16,356 | |
KDLHZDBZIXYQEI-UHFFFAOYSA-N | |
palladium | |
23938 | |
106.42 | |
Pellet |
Safety and Handling
RUO – Research Use Only