missing translation for 'onlineSavingsMsg'
Learn More

Palladium, 1% on granular carbon, reduced, Thermo Scientific Chemicals

Artikelnummer. 15412697
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
100 g
500 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

It is used in hydrogenation of aromatic and aliphatic nitro groups; in reductive alkylation/amination; hydrogenation of aromatic carbonyls, olefins, nitriles and as a catalyst in the Suzuki reaction and Stille reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 7440-05-3
Molecular Formula Pd
Molecular Weight (g/mol) 106.42
MDL Number MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745
InChI Key KDLHZDBZIXYQEI-UHFFFAOYSA-N
Synonym black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon
PubChem CID 23938
ChEBI CHEBI:33363
IUPAC Name palladium
SMILES [Pd]

Spezifikation

Physical Form Pellets
Quantity 25 g
Solubility Information Insoluble in water.
Chemical Name or Material Palladium, 1% on granular carbon, Reduced

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'