missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Palladium, 10% on activated carbon powder, eggshell, reduced, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
Description
Debenzylation. Catalysts is much easier as it greatly reduces dusting and metal loss. Used to reduce nitro compounds to amines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Physical Form | Powder |
Quantity | 50 g |
UN Number | UN3178 |
Solubility Information | Insoluble in water. |
Odor | Odorless |
Chemical Name or Material | Palladium, 10% on activated carbon powder, Eggshell, reduced |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'