missing translation for 'onlineSavingsMsg'
Learn More
Palladium, 10% on activated carbon powder, standard, reduced, nominally 50% water wet, Thermo Scientific Chemicals Product Code.: 11304417

Palladium, 10% on activated carbon powder, standard, reduced, nominally 50% water wet, Thermo Scientific Chemicals

Product Code. 11304417
100 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11304417

Brand: Thermo Scientific Alfa Aesar 038304.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Form: 900-1100m2;/g; Sold on a dry weight basis. Unit weight excludes water weight

It is a catalyst used in Stille reaction, hydrogenation reactions, Suzuki-Miyaura cross-coupling and Heck-Mizoroki reactions, deoxygenation, oxidation reactions, coupling reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-05-3
Molecular Formula Pd
Molecular Weight (g/mol) 106.42
MDL Number MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745
InChI Key KDLHZDBZIXYQEI-UHFFFAOYSA-N
Synonym black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbonShow MoreShow Less
PubChem CID 23938
ChEBI CHEBI:33363
IUPAC Name palladium
SMILES [Pd]

Specifications

Quantity 100 g
Packaging Glass bottle
Chemical Name or Material Palladium

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP31I017 Certificate TypeCertificate of Analysis Date14/04/2022 Catalog Number038304.06,038304.22,038304.14
Lot NumberU04G044 Certificate TypeCertificate of Analysis Date18/11/2021 Catalog Number038304.06,038304.22,038304.14
Lot NumberI19K20 Certificate TypeCertificate of Analysis Date18/11/2021 Catalog Number038304.06,038304.22,038304.14
Lot NumberL04T025 Certificate TypeCertificate of Analysis Date18/11/2021 Catalog Number038304.06,038304.22,038304.14
Lot NumberG26Y005 Certificate TypeCertificate of Analysis Date18/11/2021 Catalog Number038304.06,038304.22,038304.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Palladium, 10% on activated carbon powder, standard, reduced, nominally 50% water wet, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.