Learn More
Palladium on activated carbon, 10% Pd, unreduced, dry, Thermo Scientific Chemicals
31.80 € - 5870.00 €
Chemical Identifiers
CAS | 7440-05-3, 7440-44-0 |
---|---|
Molecular Formula | Pd |
Molecular Weight (g/mol) | 106.42 |
MDL Number | MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745 |
InChI Key | KDLHZDBZIXYQEI-UHFFFAOYSA-N |
Synonym | black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon |
PubChem CID | 23938 |
ChEBI | CHEBI:33363 |
IUPAC Name | palladium |
SMILES | [Pd] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10073163
|
Thermo Scientific Acros
195030010 |
1 g | Glass bottle |
31.80 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10471811
|
Thermo Scientific Acros
195030100 |
10 g | Glass Bottle |
318.00 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10687342
|
Thermo Scientific Acros
195030500 |
50 g | Glass Bottle |
1497.00 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13468438
|
Thermo Scientific Acros
195032500 |
250 g | Glass Bottle |
5870.00 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-05-3, 7440-44-0 | |
106.42 | |
KDLHZDBZIXYQEI-UHFFFAOYSA-N | |
23938 | |
palladium |
Pd | |
MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745 | |
black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon | |
CHEBI:33363 | |
[Pd] |
Specifications
Black | |
Pd | |
01,778; 04,365; 05,498; 06,443; 07,274; 15,13; 16,356 | |
black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon | |
[Pd] | |
106.42 | |
CHEBI:33363 | |
10% (Pd) | |
Powder |
7440-05-3, 7440-44-0 | |
MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745 | |
15, 7090 | |
KDLHZDBZIXYQEI-UHFFFAOYSA-N | |
palladium | |
23938 | |
106.42 | |
Particle size distribution: ≤4 Micron: 10%, ≤21 Micron: 50%, ≤90 Micron: 90% | |
Palladium on activated carbon, 10% Pd |
Safety and Handling
GHS H Statement:
Flammable solid.
GHS P Statement:
Keep container tightly closed.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning
- Palladium on activated carbon, unreduced
Signal Word
- Warning
Hazard Category
- Flammable solid Category 2
Hazard Statement
- H228-Flammable solid.
Precautionary Statement
- P233-Keep container tightly closed.
RUO – Research Use Only