missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Palladium foil, 0.25mm (0.01in) thick, 99.98+% (metals basis)
missing translation for 'orderingAttributeHoverText'
Quantity:
25 x 25 mm
50 x 50 mm
missing translation for 'unitSize'
25mm
Each
Beschreibung
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spezifikation
Spezifikation
| Quantity | 25 x 25 mm |
| Odor | Odorless |
| Weight | ≈1.9g/25x25mm |
| Form | Foil |
| Percent Purity | ≥99.98% |
| Assay | (metals basis) |
| Chemical Name or Material | Palladium foil, 0.25mm (0.01 in.) thick |
RUO – Research Use Only
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur