Learn More
Palladium hydroxide, Pd 20% on carbon, nominally 50% water, Pearlman's Catalyst, Thermo Scientific Chemicals
Description
Hydrogenolysis of benzyl-nitrogen bondsPalladium hydroxide, Pd 20% on carbon, nominally 50% water is used as a catalyst for hydrogenolysis of benzyl groups and epoxides. It is used to reduce nitro compounds to amines. It acts as a dehydrogenation reagent as well as a useful reagent for debenzylation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Used as catalyst for hydrogenolysis of benzyl groups and epoxides.
It is used to reduce nitro compounds to amines.
Acts as dehydrogenation reagent as well as useful reagent for debenzylation

Specifications
Specifications
Physical Form | Moist Powder |
Quantity | 2 g |
Solubility Information | Insoluble in water. |
Chemical Name or Material | Palladium Hydroxide, 20% on Carbon, nominally 50% water |
Safety and Handling

- Palladium hydroxide, Pd 20% on carbon, nominally 50% water, Pearlman's Catalyst
- Warning
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contains : Palladium hydroxide
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.