missing translation for 'onlineSavingsMsg'
Learn More

Palladium hydroxide, Pd 20% on carbon, nominally 50% water, Pearlman's Catalyst, Thermo Scientific Chemicals

161.81 € - 3005.96 €

Chemical Identifiers

CAS 12135-22-7
Molecular Formula H2O2Pd
Molecular Weight (g/mol) 140.43
MDL Number MFCD00064599
InChI Key NXJCBFBQEVOTOW-UHFFFAOYSA-L
Synonym palladium hydroxide, palladium ii hydroxide, pearlman's catalyst, pearlmans catalyst, dihydroxypalladium, palladiumhydroxide, pearlman catalyst, peariman's catalyst, pearl man's catalyst, pd oh 2 on carbon
PubChem CID 9942167
IUPAC Name palladium(2+);dihydroxide
SMILES [OH-].[OH-].[Pd++]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11399188
View Documents Promotion Details
Thermo Scientific Alfa Aesar
042578.04
2 g
161.81 €
2g
Please to purchase this item. Need a web account? Register with us today!
11319198
View Documents Promotion Details
Thermo Scientific Alfa Aesar
042578.09
10 g
702.50 €
10g
Please to purchase this item. Need a web account? Register with us today!
11309198
View Documents Promotion Details
Thermo Scientific Alfa Aesar
042578.18
50 g
3005.96 €
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Hydrogenolysis of benzyl-nitrogen bondsPalladium hydroxide, Pd 20% on carbon, nominally 50% water is used as a catalyst for hydrogenolysis of benzyl groups and epoxides. It is used to reduce nitro compounds to amines. It acts as a dehydrogenation reagent as well as a useful reagent for debenzylation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Used as catalyst for hydrogenolysis of benzyl groups and epoxides.
It is used to reduce nitro compounds to amines.
Acts as dehydrogenation reagent as well as useful reagent for debenzylation
Specifications

Chemical Identifiers

12135-22-7
140.43
NXJCBFBQEVOTOW-UHFFFAOYSA-L
9942167
[OH-].[OH-].[Pd++]
H2O2Pd
MFCD00064599
palladium hydroxide, palladium ii hydroxide, pearlman's catalyst, pearlmans catalyst, dihydroxypalladium, palladiumhydroxide, pearlman catalyst, peariman's catalyst, pearl man's catalyst, pd oh 2 on carbon
palladium(2+);dihydroxide

Specifications

12135-22-7
H2O2Pd
palladium hydroxide, palladium ii hydroxide, pearlman's catalyst, pearlmans catalyst, dihydroxypalladium, palladiumhydroxide, pearlman catalyst, peariman's catalyst, pearl man's catalyst, pd oh 2 on carbon
NXJCBFBQEVOTOW-UHFFFAOYSA-L
palladium(2+);dihydroxide
9942167
Moist Powder
MFCD00064599
Insoluble in water.
[OH-].[OH-].[Pd++]
140.43
Palladium Hydroxide, 20% on Carbon, nominally 50% water
Videos
Safety and Handling

Safety and Handling

P210-P240-P241-P280-P370+P378q

H228

missing translation for 'einecsNumber' : 235-219-2

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Palladium hydroxide, Pd 20% on carbon, nominally 50% water, Pearlman's Catalyst

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : Palladium hydroxide

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only