missing translation for 'onlineSavingsMsg'
Learn More

Palladium(II) acetate, 47.5% Pd

Product Code. 10790701 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
2 g
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
1g
2g
50g
This item is not returnable. View return policy

Product Code. 10790701

Brand: Thermo Scientific Acros 195180100

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3375-31-3 | C4H6O4Pd | 224.51 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3375-31-3
Molecular Formula C4H6O4Pd
Molecular Weight (g/mol) 224.51
MDL Number MFCD00012453
InChI Key YJVFFLUZDVXJQI-UHFFFAOYSA-L
Synonym palladium ii acetate, palladium diacetate, palladium acetate, diacetoxypalladium, bis acetato palladium, bisacetylpalladium, diacetatopalladium, palladium 2+ acetate, palladous acetate
PubChem CID 167845
SMILES [Pd++].CC([O-])=O.CC([O-])=O

Specifications

Melting Point 205.0°C
Quantity 10 g
Linear Formula (CH3CO2)2Pd
Merck Index 15,7092
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in acetone, benzene, methylene dichloride,, chloroform, acetonitrile and diethylether, insoluble in alcohols
IUPAC Name palladium(2+) diacetate
Formula Weight 224.51
Percent Purity 47.5% (Pd)
Packaging Glass bottle
Chemical Name or Material Palladium(II) acetate, 47.5% Pd
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.