missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Palladium(II) acetate, 47.5% Pd, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
2 g
10 g
50 g
Packaging:
Glass Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 205.0°C |
Quantity | 50 g |
Linear Formula | (CH3CO2)2Pd |
UN Number | 3261 |
Solubility Information | Solubility in water: insoluble. Other solubilities: soluble in acetone, benzene, methylene dichloride,, chloroform, acetonitrile and diethylether, insoluble in alcohols |
IUPAC Name | palladium(2+);diacetate |
Formula Weight | 224.51 |
Packaging | Glass Bottle |
Chemical Name or Material | Palladium(II) Acetate, 47.5% Pd |
Safety and Handling
Signal Word
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- Skin sensitiser Category 1
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'