Learn More
Palladium(II) acetate, Pd 45.9-48.4%, Thermo Scientific Chemicals
158.00 € - 3114.00 €
Chemical Identifiers
CAS | 3375-31-3 |
---|---|
Molecular Formula | C4H6O4Pd |
Molecular Weight (g/mol) | 224.51 |
MDL Number | MFCD00012453 |
InChI Key | YJVFFLUZDVXJQI-UHFFFAOYSA-L |
Synonym | palladium ii acetate, palladium diacetate, palladium acetate, diacetoxypalladium, bis acetato palladium, bisacetylpalladium, diacetatopalladium, palladium 2+ acetate, palladous acetate |
PubChem CID | 167845 |
IUPAC Name | palladium(2+);diacetate |
SMILES | [Pd++].CC([O-])=O.CC([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11346028
|
Thermo Scientific Alfa Aesar
010516.03 |
1 g |
158.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11356028
|
Thermo Scientific Alfa Aesar
010516.06 |
5 g |
731.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11336028
|
Thermo Scientific Alfa Aesar
010516.14 |
25 g |
3114.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Palladium(II)acetate is a catalyst of choice for a wide variety of reactions such as vinylation, Wacker process, Buchwald-Hartwig amination, carbonylation, oxidation, rearrangement of dienes (e.g., Cope rearrangement), C-C bond formation, reductive amination, etc. Precursor to Pd(0), other Pd(II) compounds of catalytic significance, and Pd nanowires.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
3375-31-3 | |
224.51 | |
YJVFFLUZDVXJQI-UHFFFAOYSA-L | |
167845 | |
[Pd++].CC([O-])=O.CC([O-])=O |
C4H6O4Pd | |
MFCD00012453 | |
palladium ii acetate, palladium diacetate, palladium acetate, diacetoxypalladium, bis acetato palladium, bisacetylpalladium, diacetatopalladium, palladium 2+ acetate, palladous acetate | |
palladium(2+);diacetate |
Specifications
∽205°C (decomposition) | |
Red to Brown | |
45.9 to 48.4% | |
MFCD00012453 | |
Soluble as monomer in glacial acetic acid or as trimer in benzene. | |
[Pd++].CC([O-])=O.CC([O-])=O | |
224.51 | |
673.46 | |
Palladium(II) acetate |
3375-31-3 | |
Needles | |
C4H6O4Pd | |
palladium ii acetate, palladium diacetate, palladium acetate, diacetoxypalladium, bis acetato palladium, bisacetylpalladium, diacetatopalladium, palladium 2+ acetate, palladous acetate | |
YJVFFLUZDVXJQI-UHFFFAOYSA-L | |
palladium(2+);diacetate | |
167845 | |
Various forms in red-brown color (powder/flake/crystalline/beads) |
Safety and Handling
Avoid contact with skin. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves and eye/face protection. This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets.
P234-P280-P305+P351+P338-P310-P390
H290-H318
missing translation for 'einecsNumber' : 222-164-4
missing translation for 'rtecsNumber' : AJ9000000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only