missing translation for 'onlineSavingsMsg'
Learn More

Palladium(II) bromide, Premion™, 99.998% (metals basis), Pd 39.5% min

Product Code. p-7058968
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11320288

Brand: Thermo Scientific Alfa Aesar 043697.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13444-94-5 | Br2Pd | 266.23 g/mol

Palladium(II) bromide is used as a source of the palladium(0) precatalyst in various coupling reactions. It is used as a catalyst for the carbonylative Heck coupling reaction. It is also used as an oxidizing agent in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13444-94-5
Molecular Formula Br2Pd
Molecular Weight (g/mol) 266.23
MDL Number MFCD00011170
InChI Key INIOZDBICVTGEO-UHFFFAOYSA-L
Synonym palladium ii bromide, palladium bromide, palladium bromide pdbr2, palladous bromide, palladium ii bromide, premion, palladium 2+ dibromide, pdbr2, palladiumbromide, palladiumbromide pdbr2
PubChem CID 83469
IUPAC Name dibromopalladium
SMILES [Br-].[Br-].[Pd++]

Specifications

Melting Point (decomposition)
Color Black
Physical Form Powder
Quantity 1 g
Solubility Information Insoluble in water. Soluble in hydrogen bromide.
Formula Weight 266.22
Percent Purity 99.998%
Grade Premion
Concentration or Composition (by Analyte or Components) Pd 39.5% min
Assay (metals basis)
Chemical Name or Material Palladium (II) bromide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.