missing translation for 'onlineSavingsMsg'
Learn More
Palladium(II) chloride, 99.9% (metals basis), Pd 59.0% min, Thermo Scientific Chemicals Product Code.: 11391658

Palladium(II) chloride, 99.9% (metals basis), Pd 59.0% min, Thermo Scientific Chemicals

Product Code. 11391658
0.5 g
Click to view available options
Quantity:
0.5 g
2 g
10 g
This item is not returnable. View return policy

Product Code. 11391658

Brand: Thermo Scientific Alfa Aesar 011034.02

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For Use As a Catalyst Precursor

Catalyst precursor. Palladium(II) chloride is used to remove the stain present in stainless steel . Its solutions are used to test the corrosion-resistance of stainless steel. It is used in carbon monoxide detectors. It is also used for cosmetic tattooing of leukomas in the cornea and in the preparation of palladium(II) iodide by reacting with potassium iodide. It is involved in the synthesis of semiconducting metal-containing polymers. It is used as a catalyst for carbonylation of organic tellurides and deamination of phenethylamines to phenyl substituted pyrroles with copper(I) as co catalyst. It plays an important role in the hydrodimerization of butadiene to form octa-2,7-dien-1-ol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-10-1
Molecular Formula Cl2Pd
Molecular Weight (g/mol) 177.32
MDL Number MFCD00003558
InChI Key PIBWKRNGBLPSSY-UHFFFAOYSA-L
PubChem CID 24290
ChEBI CHEBI:53434
SMILES [Cl-].[Cl-].[Pd++]

Specifications

Melting Point 675°C (decomposition)
Color Red to Brown
Physical Form Crystalline
Quantity 0.5 g
Assay Percent Range 99.9% (metals basis)
Synonym palladium chloride, palladium ii chloride, palladium dichloride, palladous chloride, pdcl2, palladium 2+ chloride, palladium chloride pdcl2, enplate activator 440
Solubility Information Soluble in dilute mineral acids,aqueous metal halides.Soluble in cold water,organic solvents and hydrochloric acid.
IUPAC Name dichloropalladium
Formula Weight 177.31
Odor Odorless
Appearance Red-brown powder
Density 4 g/cm3 at 20°C
Chemical Name or Material Palladium(II) chloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP01K043 Certificate TypeCertificate of Analysis Date08/03/2024 Catalog Number011034.02,011034.09,011034.18,011034.04
Lot NumberN05K050 Certificate TypeCertificate of Analysis Date15/02/2024 Catalog Number011034.02,011034.09,011034.18,011034.04
Lot NumberA0417282 Certificate TypeCertificate of Analysis Date20/07/2023 Catalog Number011034.02,011034,011034.09,011034.18,011034.04
Lot NumberY28E056 Certificate TypeCertificate of Analysis Date04/07/2023 Catalog Number011034.02,011034,011034.09,011034.18,011034.04
Lot Number10226360 Certificate TypeCertificate of Analysis Date01/05/2023 Catalog Number011034.02,011034.09,011034.18,011034.04
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Palladium(II) chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Palladium(II) chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Palladium(II) chloride, 99.9% (metals basis), Pd 59.0% min, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.