missing translation for 'onlineSavingsMsg'
Learn More

Palladium(II) chloride, 99.999%, (trace metal basis), Thermo Scientific Chemicals

200.00 € - 4057.19 €

Chemical Identifiers

CAS 7647-10-1
Molecular Formula Cl2Pd
Molecular Weight (g/mol) 177.32
MDL Number MFCD00003558
InChI Key PIBWKRNGBLPSSY-UHFFFAOYSA-L
Synonym palladium chloride, palladium ii chloride, palladium dichloride, palladous chloride, pdcl2, palladium 2+ chloride, palladium chloride pdcl2, enplate activator 440
PubChem CID 24290
ChEBI CHEBI:53434
SMILES [Cl-].[Cl-].[Pd++]
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10068881
SDS View Documents
Thermo Scientific Acros
369670010
1 g Glass bottle
200.00 €
1g
Please to purchase this item. Need a web account? Register with us today!
10542062
SDS View Documents
Thermo Scientific Acros
369670050
5 g Glass bottle
983.90 €
5g
Please to purchase this item. Need a web account? Register with us today!
11984361
SDS View Documents
Thermo Scientific Acros
369670250
25 g Glass bottle
4057.19 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
Specifications

Chemical Identifiers

7647-10-1
177.32
PIBWKRNGBLPSSY-UHFFFAOYSA-L
24290
[Cl-].[Cl-].[Pd++]
Cl2Pd
MFCD00003558
palladium chloride, palladium ii chloride, palladium dichloride, palladous chloride, pdcl2, palladium 2+ chloride, palladium chloride pdcl2, enplate activator 440
CHEBI:53434

Specifications

678.0°C to 680.0°C
White
59 to 61% (Pd)
PdCl2
15,7091
Solubility in water: 31.99g/L. Other solubilities: soluble in hydrochloric acid,hydrobromic acid,soluble in alcohol and acetone
[Cl-].[Cl-].[Pd++]
177.32
CHEBI:53434
99.999%
Palladium(II) chloride
7647-10-1
Powder
Cl2Pd
MFCD00003558
palladium chloride, palladium ii chloride, palladium dichloride, palladous chloride, pdcl2, palladium 2+ chloride, palladium chloride pdcl2, enplate activator 440
PIBWKRNGBLPSSY-UHFFFAOYSA-L
dichloropalladium
24290
177.31
Trace Metal Basis
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May be corrosive to metals.
May cause an allergic skin reaction.
Causes serious eye damage.
Harmful if swallowed.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 231-596-2

Product Identifier
  • Palladium(II) chloride

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin sensitiser Category 1

Hazard Statement
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : Palladium(II) chloride

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.