missing translation for 'onlineSavingsMsg'
Learn More

Palladium(II) iodide, 99.9% (metals basis), Pd 28% min, Thermo Scientific Chemicals

Product Code. 11321968
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Palladium(II) iodide is used in gravimetric analysis. It is involved in the synthesis of symmetrical and unsymmetrical ureas.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-38-7
Molecular Formula I2Pd
Molecular Weight (g/mol) 360.23
MDL Number MFCD00011171
InChI Key HNNUTDROYPGBMR-UHFFFAOYSA-L
IUPAC Name palladium(2+) diiodide
SMILES [Pd++].[I-].[I-]

Specifications

Melting Point 350°C (decomposition)
Color Black
Physical Form Powder
Quantity 10 g
Solubility Information Soluble in ammonia,methyl acetate and potassium iodide. Slightly soluble in hydriodic acid and concentrated nitric acid. Insoluble in water,ethanol,diluted nitric acid,hydrochloric acid and diethyl ether.
Formula Weight 360.23
Percent Purity 99.9%
Concentration or Composition (by Analyte or Components) Pd 28% min.
Density 6.003 g/mL
Assay (metals basis)
Chemical Name or Material Palladium(II) iodide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.