missing translation for 'onlineSavingsMsg'
Learn More

Palladium(II) nitrate hydrate, 99.8% (metals basis), Pd 39% min, Thermo Scientific Chemicals

Product Code. 11311668
Click to view available options
Quantity:
2 g
10 g
50 g

Palladium(II) nitrate hydrate is used as a precursor to supported palladium catalysts. It is also used as a catalyst in the conversion of alkene nitration to glycol dinitrates. It is involved in the separation of chlorine and iodine and used as biochemical for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 207596-32-5
Molecular Formula N2O6Pd
Molecular Weight (g/mol) 230.43
MDL Number MFCD00011169
InChI Key GPNDARIEYHPYAY-UHFFFAOYSA-N
Synonym palladium nitrate, palladium ii nitrate, palladous nitrate, palladium dinitrate, unii-5g27lbz05u, nitric acid, palladium 2+ salt, hydrogen tetranitropalladate ii, palladium 2+ dinitrate, palladiumdinitrate
PubChem CID 24932
IUPAC Name palladium(2+);dinitrate
SMILES [Pd++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point (decomposition)
Color Red to Brown
Physical Form Crystals
Quantity 10 g
Assay Percent Range 99.8% (metals basis)
Solubility Information Soluble in dilute nitric acid.
Formula Weight 230.43 (Anhydrous)
Sensitivity Hygroscopic
Appearance Red-brown cristals
Chemical Name or Material Palladium(II) nitrate hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.