missing translation for 'onlineSavingsMsg'
Learn More

Palladium(II) nitrate, solution, Pd 12-16% w/w (cont. Pd), Thermo Scientific™

Product Code. 11400421
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10102-05-3
Molecular Formula N2O6Pd
Molecular Weight (g/mol) 230.43
MDL Number MFCD00011169
InChI Key GPNDARIEYHPYAY-UHFFFAOYSA-N
Synonym palladium nitrate, palladium ii nitrate, palladous nitrate, palladium dinitrate, unii-5g27lbz05u, nitric acid, palladium 2+ salt, hydrogen tetranitropalladate ii, palladium 2+ dinitrate, palladiumdinitrate
PubChem CID 24932
IUPAC Name palladium(2+);dinitrate
SMILES [Pd++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Color Brown to Orange
UN Number UN3264
Concentration Pd 12 to 16% w/w (cont. Pd)
Solubility Information Soluble in water,nitric acid.
Formula Weight 230.43
Quantity 1 g
Physical Form Liquid
Chemical Name or Material Palladium(II) nitrate, Solution
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'