missing translation for 'onlineSavingsMsg'
Learn More

Palladium(II) oxide, anhydrous, 99.9% (metals basis), Pd 85% min

Product Code. p-7048620
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11321678

Brand: Thermo Scientific Alfa Aesar 011040.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Catalytic reduction, in organic synthesis | CAS: 1314-08-5 | OPd | 122.42 g/mol

Catalytic reduction, in organic synthesis.Palladium(II) oxide is used as a catalyst for reduction in organic synthesis. It is an excellent gas sensing material for carbon monoxide at low working temperatures. It is also used in the preparation of good optical quality films. It is involved in the preparation of LuPd2O4 , which is used in superconducting materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-08-5
Molecular Formula OPd
Molecular Weight (g/mol) 122.42
MDL Number MFCD00011172
InChI Key JQPTYAILLJKUCY-UHFFFAOYSA-N
Synonym palladium ii oxide, palladium oxide pdo, acmc-1bqvh, palladium oxide, pdo, palladium ii oxide, anhydrous, palladium ii oxide-pd, 11113-77-2 cpd with unspecified mf, palladium oxide ? inverted exclamation mark-?, palladium ii oxide trace metals basis
PubChem CID 5083724
SMILES [O--].[Pd++]

Specifications

Melting Point 870°C
Color Brown
Physical Form Powder
Quantity 1 g
Merck Index 14,6993
Solubility Information Slightly soluble in aqua regia; Insoluble in water and acids.
IUPAC Name oxopalladium
Formula Weight 122.4
Percent Purity 99.9%
Concentration or Composition (by Analyte or Components) Pd 85% min.
Odor Odorless
Density 8.70 g/mL
Chemical Name or Material Palladium(II) oxide, Anhydrous
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.