missing translation for 'onlineSavingsMsg'
Learn More

Palladium(II) sulfate dihydrate, Premion™, 99.995% (metals basis), Thermo Scientific Chemicals

Product Code. 11340288
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13444-98-9
Molecular Formula H4O6PdS
Molecular Weight (g/mol) 238.51
MDL Number MFCD00011173
InChI Key TWIRRPLUAGEFNJ-UHFFFAOYSA-L
Synonym palladium ii sulfate dihydrate, palladium ii sulfate dihydrate, premion, palladium sulfate dihydrate, so4.pd.2h2o, palladium 2+ sulfate dihydrate, palladium 2+ dihydrate sulfate, palladium 2+ sulfate-water 1/1/2, palladium 2+ ion dihydrate sulfate, palladium ii sulfate dihydrate metals basis
PubChem CID 22042048
IUPAC Name palladium(2+);sulfate;dihydrate
SMILES O.O.[Pd++].[O-]S([O-])(=O)=O

Specifications

Melting Point (decomposition)
Color Brown to Red
Physical Form Powder
Quantity 1 g
Linear Formula PdSO4·2H2O
UN Number UN3260
Formula Weight 238.51 (202.48 Anhydrous)
Percent Purity 99.995%
Grade Premion™
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Palladium (II) sulfate dihydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.