missing translation for 'onlineSavingsMsg'
Learn More

Palladium(II) sulfide, 99% (metals basis), Pd 72% min, Thermo Scientific Chemicals

Product Code. 11307507
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is used in scientific research and development.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12125-22-3
Molecular Formula PdS
Molecular Weight (g/mol) 138.48
MDL Number MFCD00016285
InChI Key FEWBURNPAYHODO-UHFFFAOYSA-N
Synonym palladium ii sulfide, palladium sulphide, palladium monosulphide, palladium sulfide, acmc-20ajzj, palladium sulfide,, palladium ii sulfide trace metals basis
PubChem CID 82926
SMILES [S--].[Pd++]

Specifications

Melting Point 950°C (decomposition)
Physical Form Crystalline
Quantity 1 g
Solubility Information Insoluble in water.
IUPAC Name sulfanylidenepalladium
Formula Weight 138.46
Percent Purity 99%
Concentration or Composition (by Analyte or Components) Pd 72% min.
Odor Sulphurous
Density 6.60 g/mL
Assay (metals basis)
Chemical Name or Material Palladium(II) sulfide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'