missing translation for 'onlineSavingsMsg'
Learn More

Palladium(II) trimethylacetate, Thermo Scientific Chemicals

Product Code. 15442747
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

 

Palladium(II) trimethylacetate is used as a catalyst for aerobic cyclization of acrylic acid with alkenes. It is utilized in the benzylation of heterocycles with benzyl chloride and direct arylation reactions. Furthermore, it is used in regioselective oxidative arene cross-coupling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 230°C to 232°C
CAS 106224-36-6
Color Orange
Physical Form Solid
Quantity 1 g
Molecular Formula C10H18O4Pd
Linear Formula [(CH3)3CCO2]2Pd
MDL Number MFCD16038138
Synonym palladium pivalate, palladium ii pivalate, palladium ii trimethylacetate, acmc-20ajzo, palladium 11 pivalate, palladium 2+ dipivalate, dipivalic acid palladium ii salt, palladium 2+ ion dipivalate, palladium 2+ bis 2,2-dimethylpropanoate
Solubility Information Soluble in ethyl acetate,methanol,chloroform,tetrahydrofuran and 1,4-dioxane. Insoluble in water.
InChI Key PAGZTSLSNQZYEV-UHFFFAOYSA-L
SMILES CC(C)(C)C(=O)[O-].CC(C)(C)C(=O)[O-].[Pd+2]
IUPAC Name 2,2-dimethylpropanoate;palladium(2+)
Molecular Weight (g/mol) 308.67
PubChem CID 24905427
Formula Weight 308.67
Chemical Name or Material Palladium(II) trimethylacetate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'